MilliporeSigma
  • Bicelle membranes and their use for hydrophobic peptide studies by circular dichroism and solid state NMR.
Bicelle membranes and their use for hydrophobic peptide studies by circular dichroism and solid state NMR.
Biochimica et biophysica acta (2005-06-18)
Cécile Loudet, Lucie Khemtémourian, Fabien Aussenac, Stéphane Gineste, Marie-France Achard, Erick J Dufourc
ABSTRACT
Mixtures of dicaproyl- (DC), dimyristoyl- (DM) and 1-tetradecanoyl-2-biphenylbutanoyl-(TBB) phosphatidylcholine (PC) in water produce bicelle membranes that are oriented by magnetic fields. DMPC/DCPC systems orient such that their membrane plane is parallel to the magnetic field, whereas for TBBPC/DCPC, the plane is perpendicular to the field. Partial temperature-composition-hydration diagrams are established using solid-state 31P-NMR. DMPC/DCPC bicelles exist on a large range of composition but on a narrow temperature domain (25-45 degrees C). At converse, TBBPC/DCPC form bicelles on a narrow compositional range but over a large temperature span (10-70 degrees C). The TBBPC/DCPC bicelles are shown to be a very powerful potential tool to study the orientation of hydrophobic helices in membranes using wide line 15N-NMR. The DMPC/DCPC system that undergoes a micelle-to-bicelle transition on going from 10 degrees C to 40 degrees C may be used with circular dichroism to study the state of association of hydrophobic helices within the membrane. Results suggest that the transmembrane fragment of the neu/erbB-2 receptor is monomeric in micellar medium and dimeric/multimeric in bicelle membranes.