MilliporeSigma
1,3-Dimethyllumazine derivatives from Limnatis nilotica.
Journal of natural products (2005-06-25)
Gerard Voerman, Silvia Cavalli, Gijsbert A van der Marel, Wolfgang Pfleiderer, Jacques H van Boom, Dmitri V Filippov
ABSTRACT
Two previously unknown lumazine derivatives, 1 and 2, have been isolated from the parasitic freshwater leech Limnatis nilotica. The structures of the compounds have been elucidated by NMR and unambiguously corroborated by chemical synthesis.