MilliporeSigma
  • Antioxidant activity of melatonin and a pinoline derivative on linoleate model system.
Antioxidant activity of melatonin and a pinoline derivative on linoleate model system.
Journal of pineal research (2005-06-28)
J Mekhloufi, D Bonnefont-Rousselot, S Yous, D Lesieur, M Couturier, P Thérond, A Legrand, D Jore, M Gardès-Albert
ABSTRACT
This study aimed at investigating the in vitro protective effects of GWC22, a novel pinoline derivative [6-ethyl-1-(3-methoxyphenyl)-2-propyl-1,2,3,4-tetrahydro-beta-carboline] chlorhydrate, against radiation-induced oxidation of linoleate initiated by hydroxyl radicals ((*)OH). Using linoleate micelles (10(-2) m) as lipid model, two indexes of peroxidation have been measured, i.e. conjugated dienes and hydroperoxides. Similar determinations were performed with melatonin in order to compare the protective effects of the two compounds. It was observed that, the higher the concentration of GWC22 (or melatonin) (3 x 10(-5) to 10(-4) m), the stronger the antioxidant ability. In these in vitro assays, GWC22 showed a better antioxidant effect than melatonin for a given antioxidant concentration. A reaction scheme has been proposed to explain the inhibitory effect of an antioxidant via the propagating steps of the lipid peroxidation. Indeed, we have suggested that melatonin and GWC22 may compete with the fatty acid to scavenge lipid peroxyl radicals (LOO(*)). We have estimated a lower limit for the LOO(*) rate constant for GWC22 (>/=1.4 x 10(5)/m/s) and for melatonin (>/=2.8 x 10(4)/m/s) assuming that the k-value of the propagating step in linoleate (LOO(*) + linoleate) was 1.4 x 10(3)/m/s. The difference of reactivity between melatonin and GWC22 in this model system is assumed to be related to their relative lipophilicity.