MilliporeSigma
  • Synthesis of morphiceptin (Tyr-Pro-Phe-Pro-NH(2)) by dipeptidyl aminopeptidase IV derived from Aspergillus oryzae.
Synthesis of morphiceptin (Tyr-Pro-Phe-Pro-NH(2)) by dipeptidyl aminopeptidase IV derived from Aspergillus oryzae.
Journal of agricultural and food chemistry (2005-07-21)
Toru Ota, Aki Itoh, Hiroshi Tachi, Keita Kudoh, Tatsuo Watanabe, Yuji Yamamoto, Tadahiro Tadokoro, Akio Maekawa
ABSTRACT
Morphiceptin (Tyr-Pro-Phe-Pro-NH(2)), tetrapeptide, was synthesized using dipeptidyl aminopeptidase IV (DP IV, EC 3.4.14.5) derived from Aspergillus oryzae RIB 915 as a catalyst. Tyr-Pro-OEt was incubated with Phe-Pro-NH(2) in the presence of DP IV under various conditions of temperature, concentrations of ethylene glycol, pH, reaction time, and others. Morphiceptin was obtained at 40% yield under the optimal reaction conditions: substrate, 4 mM Tyr-Pro-OEt.HCl and 20 mM Phe-Pro-NH(2).HCl; enzyme, DP IV, 0.275 nkat; solvent, 60% ethylene glycol containing 20 mM phosphate buffer at pH 7.0; amine, 4.2 mM diisopropylamine at 4 degrees C for 24 h. Amino group protection was unnecessary for synthesis of morphiceptin by DP IV.