MilliporeSigma
  • Kinetic study of cytochrome P450 3A4 activity on warfarin by capillary electrophoresis with fluorescence detection.
Kinetic study of cytochrome P450 3A4 activity on warfarin by capillary electrophoresis with fluorescence detection.
Journal of chromatography. A (2005-07-23)
Jie Zhang, Pham Thi Thanh Ha, Yijia Lou, Jos a Hoogmartens, Ann Van Schepdael
ABSTRACT
The use of capillary electrophoresis (CE) for the determination of cytochrome P450 3A4 (CYP3A4) activity with R-warfarin as a substrate was investigated. CYP3A4 activity was determined by the quantitation of the product, 10-hydroxywarfarin, based on separation by CE. The separation conditions were as follows: capillary, 80.5 cm (75 microm i.d., 60 cm effective length); 50 mM sodium phosphate buffer (pH 6.5); 23 kV (90 microA) applied voltage; fluorescence detection, excitation wavelength, 310 nm, emission wavelength, 418 nm; capillary temperature, 37 degrees C. With the developed CYP3A4 activity assay and the Lineweaver-Burk equation, the Michaelis-Menten parameters Km and Vmax for formation of 10-hydroxywarfarin from R-warfarin in the presence of CYP3A4 were calculated to be 166 +/- 12 microM and 713 +/- 14 pmol/min/nmol (or 91.4 pmol/min/mg) CYP3A4, respectively.