MilliporeSigma
  • [The mechanism of acetate assimilation in purple nonsulfur bacteria lacking the glyoxylate pathway: acetate assimilation in Rhodobacter sphaeroides cells].
[The mechanism of acetate assimilation in purple nonsulfur bacteria lacking the glyoxylate pathway: acetate assimilation in Rhodobacter sphaeroides cells].
Mikrobiologiia (2005-08-27)
L V Filatova, I A Berg, E N Krasil'nikova, A A Tsygankov, T V Laurinavichene, R N Ivanovskiĭ
ABSTRACT
The mechanism of acetate assimilation in the purple nonsulfur bacterium Rhodobacter sphaeroides, which lacks the glyoxylate pathway, is studied. It is found that the growth of this bacterium in batch and continuous cultures and the assimilation of acetate in cell suspensions are not stimulated by bicarbonate. The consumption of acetate is accompanied by the excretion of glyoxylate and pyruvate into the medium, stimulated by glyoxylate and pyruvate, and inhibited by citramalate. The respiration of cells in the presence of acetate is stimulated by glyoxylate, pyruvate, citramalate, and mesaconate. These data suggest that the citramalate cycle may function in Rba. sphaeroides in the form of an anaplerotic pathway instead of the glyoxylate pathway. At the same time, the low ratio of fixation rates for bicarbonate and acetate exhibited by the Rba. sphaeroides cells (approximately 0.1), as well as the absence of the stimulatory effect of acetate on the fixation of bicarbonate in the presence of the Calvin cycle inhibitor iodoacetate, suggests that pyruvate synthase is not involved in acetate assimilation in the bacterium Rba. sphaeroides.