MilliporeSigma
  • Synthesis and reactions of 2-chloro- and 2-tosyloxy-2'-deoxyinosine derivatives.
Synthesis and reactions of 2-chloro- and 2-tosyloxy-2'-deoxyinosine derivatives.
The Journal of organic chemistry (2005-08-27)
Narender Pottabathini, Suyeal Bae, Padmanava Pradhan, Hoh-Gyu Hahn, Heduck Mah, Mahesh K Lakshman
ABSTRACT
Convenient syntheses of 2-chloro- and 2-tosyloxy-2'-deoxyinosine as their tert-butyldimethylsilyl ethers are described. Both compounds can be synthesized via a common route and rely on commercially available 2'-deoxyguanosine. The present method leading to the chloro nucleoside is operationally simpler compared to previously reported glycosylation techniques where isomeric products were obtained. Both electrophilic nucleosides can be used for the preparation of N-substituted 2'-deoxyguanosine analogues via displacement of the leaving groups, and a comparison of their reactivities shows the chloro analogue to be superior. Interestingly, a Pd catalyst-mediated, two-step, one-pot conversion of an allyl-protected chloro nucleoside intermediate to the final modified 2'-deoxyguanosine derivatives is also feasible. On the basis of these observations, initial assessments of Pd-catalyzed aryl amination as well as a C-C cross-coupling have also been performed with the chloro and tosyloxy nucleoside substrates. Results indicate a potentially high synthetic utility of 2-chloro-2'-deoxyinosine and in many instances this derivative can supplant the bromo and fluoro analogues that are more cumbersome to prepare or are not readily available.
Sigma-Aldrich
2′-Deoxyinosine, ≥98%