MilliporeSigma
  • The combination of sulfinosine and 8-Cl-cAMP induces synergistic cell growth inhibition of the human neuroblastoma cell line in vitro.
The combination of sulfinosine and 8-Cl-cAMP induces synergistic cell growth inhibition of the human neuroblastoma cell line in vitro.
Investigational new drugs (2005-12-28)
Dragana Janković, Milica Pesić, Jasna Marković, Selma Kanazir, Ivanka Marković, Ljubisav Rakić, Sabera Ruzdijić
ABSTRACT
To identify purine analogs that could be effective in treating neuroblastomas, we tested the anticancer properties of sulfinosine, 8-Cl-cAMP and 8-Cl-adenosine in the SK-N-SH cell line. First we examined the effects of these three agents on cell growth inhibition and cell viability by the BrdU and Sulforhodamine B assay. Treatment of SK-N-SH cells with increasing concentrations of these compounds led to a significant inhibition of cell proliferation and decrease of cell viability in a time- and dose-dependent manner at micromolar concentration (<10 microm). Treatment with a combination of sulfinosine and 8-Cl-cAMP resulted in synergistic effects on growth inhibition, cell cycle arrest and induction of apoptosis. Flow-cytometric analysis showed that 8-Cl-cAMP arrested the cells in the G0/G1 phase and sulfinosine blocked cell cycle progression at the G2/M stage, in contrast to the combined effects of both agents that did not arrest growth at any particular phase of the cell cycle. Further analysis of apoptosis induction demonstrated an increase from 17 to 24% of both early and late apoptotic cells and a very low percentage of necrotic cells. These results indicate that apoptosis was the predominant type of cell death after treatment of SK-N-SH cells with both substances, as well as with their combinations.
Sigma-Aldrich
8-Chloroadenosine 3′,5′-cyclic-monophosphate, ≥85%