MilliporeSigma
  • Evaluation of rapidly disintegrating tablets containing glycine and carboxymethylcellulose.
Evaluation of rapidly disintegrating tablets containing glycine and carboxymethylcellulose.
International journal of pharmaceutics (2006-01-26)
Jinichi Fukami, Etsuo Yonemochi, Yasuo Yoshihashi, Katsuhide Terada
ABSTRACT
A rapidly disintegration tablet in the oral cavity was prepared using a glycine as a disintegrant. Effect of disintegrant on the disintegration behavior of the tablet in the oral cavity was evaluated. Wetting time prepared from carboxymethylcellulose (NS-300) having the hardness of 4 kg was 3 s. Tablet containing NS-300 showed fastest disintegration compared to other formulations. These results suggested that NS-300 possessed excellent wetting nature and resulted in the rapid disintegration of tablet. Ethenzamide and ascorbic acid were added to the formulation, and their disintegration behavior were evaluated. Ethenzamide did not affect the disintegration property, however, ascorbic acid prolonged disintegration time. It was suggested that the tablet formulation containing NS-300 and glycine was highly applicable to water-insoluble drug, such as ethenzamide.
Sigma-Aldrich
2-Ethoxybenzamide, 97%