MilliporeSigma
  • In vivo mutagenesis in the lungs of gpt-delta transgenic mice treated intratracheally with 1,6-dinitropyrene.
In vivo mutagenesis in the lungs of gpt-delta transgenic mice treated intratracheally with 1,6-dinitropyrene.
Environmental and molecular mutagenesis (2006-02-21)
Akiko H Hashimoto, Kimiko Amanuma, Kyoko Hiyoshi, Hirohisa Takano, Ken-ichi Masumura, Takehiko Nohmi, Yasunobu Aoki
ABSTRACT
1,6-Dinitropyrene (1,6-DNP) is a ubiquitous airborne pollutant found in diesel exhaust. In this study, mutagenesis was examined in the lungs of gpt-delta transgenic mice after intratracheal instillation of 0-0.1 mg 1,6-DNP. In addition, the 1,6-DNP-induced gpt mutation spectrum was compared with that of control mice. A single intratracheal injection of 0-0.05 mg 1,6-DNP resulted in significant dose-dependent increases in mutant frequency; the induced mutant frequency declined at the 0.1 mg dose. The average lung mutant frequencies at doses of 0.025, 0.05, and 0.1 mg 1,6-DNP were 2.9-, 4.1-, and 1.9-times higher than for control mice ((0.50+/-0.16)x10(-5)). The major mutations induced by 1,6-DNP included G:C-->A:T transitions, G:C-->T:A transversions, and 1-base deletions. Among the G:C-->A:T transitions isolated from 1,6-DNP-treated mice, five (at nucleotide positions 64, 110, 115, 116, and 418) were observed in four or more animals. These positions therefore are potential hotspots for 1,6-DNP mutation. The predominant frameshift mutations following 1,6-DNP treatment included single base pair deletions at G:C (9/13=69%). The results of this study indicate that 1,6-DNP is mutagenic for the lungs of mice.