MilliporeSigma
  • Phencyclidine-induced cognitive deficits in mice are improved by subsequent subchronic administration of fluvoxamine: role of sigma-1 receptors.
Phencyclidine-induced cognitive deficits in mice are improved by subsequent subchronic administration of fluvoxamine: role of sigma-1 receptors.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2006-02-24)
Kenji Hashimoto, Yuko Fujita, Masaomi Iyo
ABSTRACT
This study was undertaken to examine the effects of the selective serotonin reuptake inhibitors fluvoxamine and paroxetine on cognitive deficits in mice after repeated administration of the N-methyl-D-aspartate receptor antagonist phencyclidine (PCP). In the novel object recognition test, repeated administration of PCP (10 mg/kg/day, 10 days) significantly decreased the exploratory preference in the retention test session, but not in the training test session. PCP-induced cognitive deficits were significantly improved by subsequent subchronic (2-week) administration of fluvoxamine (20 mg/kg/day), but not paroxetine (10 mg/kg/day). Furthermore, the effect of fluvoxamine on PCP-induced cognitive deficits was antagonized by co-administration of the selective sigma-1 receptor antagonist NE-100 (1 mg/kg/day). Moreover, PCP-induced cognitive deficits were also significantly improved by subsequent subchronic (2-week) administration of the selective sigma-1 receptor agonist SA4503 (1 mg/kg/day) or neurosteroid dehydroepiandrosterone 3-sulfate (DHEA-S; 25 mg/kg/day). The effects of SA4503 or DHEA-S were also antagonized by co-administration of NE-100 (1 mg/kg/day), suggesting the role of sigma-1 receptors in the active mechanisms of these drugs. In contrast, acute single administration of these drugs (fluvoxamine, paroxetine, SA4503) alone or combination with NE-100 did not alter PCP-induced cognitive deficits. The present study suggests that agonistic activity of fluvoxamine at sigma-1 receptors plays a role in the active mechanisms of fluvoxamine on PCP-induced cognitive deficits in mice. Therefore, sigma-1 receptor agonists such as fluvoxamine would be potential therapeutic drugs for the treatment of the cognitive deficits of schizophrenia.