MilliporeSigma
  • Chaetoglobosin U, a cytochalasan alkaloid from endophytic Chaetomium globosum IFB-E019.
Chaetoglobosin U, a cytochalasan alkaloid from endophytic Chaetomium globosum IFB-E019.
Journal of natural products (2006-02-28)
Gang Ding, Yong C Song, Jing R Chen, Chen Xu, Hui M Ge, Xiao T Wang, Ren X Tan
ABSTRACT
A new cytotoxic cytochalasan-based alkaloid named chaetoglobosin U (1), along with four known analogues, chaetoglobosins C (2), F (3), and E (5) and penochalasin A (4), has been characterized from the EtOAc extract of a solid culture of Chaetomium globosum IFB-E019, an endophytic fungus residing inside the stem of healthy Imperata cylindrica. The structure of chaetoglobosin U was determined through correlative analyses of its UV, IR, CD, MS, and 1D ((1)H and(13)C NMR and DEPT) and 2D NMR (COSY, NOESY, HMQC, and HMBC) data. Chaetoglobosin U (1) exhibited cytotoxic activity against the human nasopharyngeal epidermoid tumor KB cell line with an IC(50) value of 16.0 microM, comparable to that (14.0 microM) of 5-fluorouracil co-assayed as a positive reference. The known analogues 2-5 were moderately active to the cell line, with IC(50) values of 34.0, 52.0, 48.0, and 40.0 microM, respectively.