MilliporeSigma
  • Peripheral interstitial keratitis: a novel manifestation of ocular mastocytosis.
Peripheral interstitial keratitis: a novel manifestation of ocular mastocytosis.
Cornea (2006-04-25)
M Teresa Magone, Irina Maric, David G Hwang
ABSTRACT
To report a case of peripheral interstitial keratitis in a patient with mastocytosis. Clinical case description and immunohistologic examination of biopsied ocular tissue. A 22-year-old woman with biopsy-proven urticaria pigmentosa, a subset of mastocytosis, presented with an active peripheral interstitial keratitis with vascularization associated with foreign body sensation and itching. Biomicroscopy of the cornea showed deep corneal inflammatory infiltrates and midstromal vascularization adjacent to a region of superior bulbar conjunctiva, which was mildly chemotic and inflamed. Topical mast cell stabilizers and a short course of topical steroids produced dramatic resolution of the lesion. Biopsy of the erythematous conjunctiva adjacent to the area of corneal inflammation showed the presence of mast cells. This is the first case of corneal inflammatory infiltration in a patient with mastocytosis. Therapy for this condition consists of a combination of topical mast cell stabilizers, topical steroids, and systemic antihistaminic therapy.