MilliporeSigma
  • Signaling mechanisms of down-regulation of voltage-activated Ca2+ channels by transient receptor potential vanilloid type 1 stimulation with olvanil in primary sensory neurons.
Signaling mechanisms of down-regulation of voltage-activated Ca2+ channels by transient receptor potential vanilloid type 1 stimulation with olvanil in primary sensory neurons.
Neuroscience (2006-05-09)
Z-Z Wu, S-R Chen, H-L Pan
ABSTRACT
Olvanil ((N-vanillyl)-9-oleamide), a non-pungent transient receptor potential vanilloid type 1 agonist, desensitizes nociceptors and alleviates pain. But its molecular targets and signaling mechanisms are little known. Calcium influx through voltage-activated Ca(2+) channels plays an important role in neurotransmitter release and synaptic transmission. Here we determined the effect of olvanil on voltage-activated Ca(2+) channel currents and the signaling pathways in primary sensory neurons. Whole-cell voltage-clamp recordings were performed in acutely isolated rat dorsal root ganglion neurons. Olvanil (1 microM) elicited a delayed but sustained inward current, and caused a profound inhibition (approximately 60%) of N-, P/Q-, L-, and R-type voltage-activated Ca(2+) channel current. Pretreatment with a specific transient receptor potential vanilloid type 1 antagonist or intracellular application of 1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid abolished the inhibitory effect of olvanil on voltage-activated Ca(2+) channel current. Calmodulin antagonists (ophiobolin-A and calmodulin inhibitory peptide) largely blocked the effect of olvanil and capsaicin on voltage-activated Ca(2+) channel current. Furthermore, calcineurin (protein phosphatase 2B) inhibitors (deltamethrin and FK-506) eliminated the effect of olvanil on voltage-activated Ca(2+) channel current. Notably, 1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid, calmodulin antagonists, and calcineurin inhibitors each alone significantly increased the amplitude of voltage-activated Ca(2+) channel current. In addition, double immunofluorescence labeling revealed that olvanil induced a rapid internalization of Ca(V)2.2 immunoreactivity from the membrane surface of dorsal root ganglion neurons. Collectively, this study suggests that stimulation of non-pungent transient receptor potential vanilloid type 1 inhibits voltage-activated Ca(2+) channels through a biochemical pathway involving intracellular Ca(2+)-calmodulin and calcineurin in nociceptive neurons. This new information is important for our understanding of the signaling mechanisms of desensitization of nociceptors by transient receptor potential vanilloid type 1 analogues and the feedback regulation of intracellular Ca(2+) and voltage-activated Ca(2+) channels in nociceptive sensory neurons.
Sigma-Aldrich
Olvanil, powder