MilliporeSigma
  • Determination of Kamlet-Taft solvent parameters pi* of high pressure and supercritical water by the UV-Vis absorption spectral shift of 4-nitroanisole.
Determination of Kamlet-Taft solvent parameters pi* of high pressure and supercritical water by the UV-Vis absorption spectral shift of 4-nitroanisole.
Physical chemistry chemical physics : PCCP (2006-05-12)
Kimitaka Minami, Masamichi Mizuta, Muneyuki Suzuki, Takafumi Aizawa, Kunio Arai
ABSTRACT
Kamlet-Taft solvent parameters, pi*, of high pressure and supercritical water were determined from 16-420 degrees C based on solvatochromic measurements of 4-nitroanisole. For the measurements, an optical cell that could be used at high temperatures and pressures was developed with the specification of minimal dead space. The low dead space cell allowed us to measure the absorption spectra of 4-nitroanisole at high temperature conditions before appreciable decomposition occurred. The behavior of pi* in terms of water density (pi* = 1.77rho- 0.71) was found to be linear, except in the near critical region, in which deviations were observed that could be attributed to local density augmentation. Excess density, which was defined as the difference between local density and bulk density, showed a maximum near the critical density of water. The frequencies of UV-Vis spectra of 4-(dimethylamino)benzonitrile and N,N-dimethyl-4-nitroaniline were correlated with pi* based on a linear solvation energy relationship (LSER) theory. Local density augmentation around 4-nitroanisole and that around 4-(dimethylamino)benzonitrile were similar but the augmentation observed around N,N-dimethyl-4-nitroaniline was larger.
Sigma-Aldrich
4-Nitroanisole, 97%