MilliporeSigma
  • Induction of wheat and maize glutathione S-transferase by some herbicide safeners and their effect on enzyme activity against butachlor and terbuthylazine.
Induction of wheat and maize glutathione S-transferase by some herbicide safeners and their effect on enzyme activity against butachlor and terbuthylazine.
Pest management science (2006-07-13)
Luciano Scarponi, Elisa Quagliarini, Daniele Del Buono
ABSTRACT
The expression of glutathione S-transferase (GST) activity in wheat and maize shoots was investigated in response to treatments with the herbicide safeners benoxacor, cloquintocet-mexyl, fenchlorazole-ethyl, fenclorim, fluxofenim and oxabetrinil. These safeners significantly enhanced the GST activity towards 1-chloro-2,4-dinitrobenzene (CDNB) as a 'standard' substrate, with the exception of oxabetrinil in maize. The enhancements of GST (CDNB) activity were found to be concomitant with increases in V(max) (the reaction rate when the enzyme is fully saturated by the substrate) in wheat following cloquintocet-mexyl and fenchlorazole-ethyl treatments, and in maize following fenchlorazole-ethyl treatment. Otherwise, decreases in V(max) were observed in wheat and maize following fenclorim and fluxofenim treatments. With the exception of oxabetrinil, all the safeners significantly reduced the apparent K(M) (the substrate concentration required for 50% of maximum GST activity) of both wheat and maize GST. The V(max) and K(M) variations following safener treatments are discussed in terms of an increased expression of GST enzymes and an increased affinity for the CDNB substrate. The activity of wheat and maize GST was also assayed towards butachlor and terbuthylazine respectively; the results indicate the ability of cloquintocet-mexyl, fenchlorazole-ethyl and fluxofenim to enhance the enzyme activity in wheat and of benoxacor and fenchlorazole-ethyl to do so in maize.
Product Description
Supelco
Benoxacor, PESTANAL®, analytical standard