MilliporeSigma
  • Evidence against alpha-adrenoceptors mediating relaxation in rat thoracic aortae: alpha-agonists relaxation depends on interaction with alpha-adrenoceptors.
Evidence against alpha-adrenoceptors mediating relaxation in rat thoracic aortae: alpha-agonists relaxation depends on interaction with alpha-adrenoceptors.
Fundamental & clinical pharmacology (2006-07-27)
Enrique F Castillo, Cindy S Ortíz, Ruth M López, Antonio Ruíz, Juan M Vélez, Carlos Castillo
ABSTRACT
In rat aorta, the presence of functional alpha(2)-adrenoceptors (alpha(2)-AR) was investigated in ring preparations preconstricted with alpha(1)-adrenergic and non- alpha(1)-adrenergic agonists. Particularly, the hypothetical interference of alpha(2)-AR agonists with alpha(1)-AR-mediated vasoconstriction was evaluated. Relaxant and contractile responses to alpha(2)-AR agonists were obtained. In endothelium-intact and endothelium-denuded aortic rings preconstricted with phenylephrine (1 x 10(-6) m), the imidazoline derivatives, clonidine and UK14304, induced relaxations with similar order of potencies (-log EC(50)) and maxima relaxant effects respectively. Pretreatment with the NO synthase inhibitor, N(G)-nitro-L-arginine methyl ester (L-NAME) had no effect on the relaxant responses to clonidine and UK14304. In phenylephrine-constricted rings with endothelium, relaxations to clonidine and UK 14304 were not antagonized by the selective alpha(2)-AR antagonist, rauwolscine (< or =1 x 10(-6) m). Clonidine and UK 14304 induced only contractions on endothelium-intact and endothelium-denuded aortic rings contracted with prostaglandin F(2alpha) (3 x 10(-7) m). Moreover, clonidine and UK 14304-induced relaxation of endothelium-denuded arteries precontracted with methoxamine but not with serotonin. Finally, the concentration-contraction curves to clonidine and UK 14304 in endothelium-denuded aortic rings were significantly shifted to the right by the alpha(1D)-AR selective antagonist, BMY 7378, and rauwolscine. The pA(2) and pK(B) values for BMY 7378 and rauwolscine, respectively, against endothelium-independent actions of clonidine and UK 14304 were characteristic of an effect on the alpha(1D)-AR. The other selective alpha(2)-AR agonist tested BHT 933 (an azepine derivative), lacks considerable relaxant and contractile effects in rat aorta. The results provide no evidence for the presence of functional alpha(2)-AR in rat aorta. Respectively, the relaxant and contractile effects of the imidazoline derivatives, clonidine and UK 14304, may be due to an adjustable (in relation to the agonist-dependent active state of the alpha(1)-AR), inhibitory and excitatory, interaction with alpha(1)-ARs.