MilliporeSigma
  • X-ray crystal structure of phenylglycine hydrazide: synthesis and spectroscopic studies of its transition metal complexes.
X-ray crystal structure of phenylglycine hydrazide: synthesis and spectroscopic studies of its transition metal complexes.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-09-05)
Kalagouda B Gudasi, Manjula S Patil, Ramesh S Vadavi, Rashmi V Shenoy, Siddappa A Patil, M Nethaji
ABSTRACT
Phenylglycine hydrazide was synthesized and investigated by X-ray crystallography. It crystallizes in the monoclinic space group P121/c with cell parameters a=5.9459 (18) Angstrom, b=5.1940 (16) Angstrom, c=26.7793 (83) Angstrom and Z=2. Its conformational changes, on complexation with transition metal ions Cu(II), Co(II), Ni(II), Mn(II) and Zn(II) has been studied on the basis of elemental analysis, magnetic moment and spectral (IR, (1)H NMR, UV-vis) studies. The bidentate nature of the ligand was confirmed on the basis of a comparative IR and NMR spectral studies. The trigonal bipyramidal geometries were observed for Cu(II), Ni(II) and Co(II) complexes, while it is octahedral for the remaining complexes. The conductivity data suggest them to be non-electrolytes.