MilliporeSigma
  • A cooperative beads-on-a-string approach to exceptionally stable DNA triplexes.
A cooperative beads-on-a-string approach to exceptionally stable DNA triplexes.
Chemical communications (Cambridge, England) (2006-09-14)
Yan Zheng, Hai Long, George C Schatz, Frederick D Lewis
ABSTRACT
A poly(dT) oligomer can serve as the string on which synthetic hairpins possessing poly(dA) and poly(dT) arms connected by a hydrophobic perylene diimide linker are assembled like beads on a string. The synthetic hairpins form head-to-head dimers and trimers, respectively, in the absence of the poly(dT) string and in the presence of a string with inverted polarity at mid-strand. However, they assemble in cooperative head-to-tail fashion on normal poly(dT) oligomers.