MilliporeSigma
[Studies on the chemical constituents in herb of Ranunculus japonicus].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2006-10-20)
Wei Zheng, Chang-xin Zhou, Shui-li Zhang, Lin-jia Weng, Yu Zhao
ABSTRACT
To isolate and determine the chemical constituents from Ranunculus japonicus. Compounds were isolated by column chromatography, and identified by MS, NMR data. Nine compounds were obtained andelucidated as scoparone (1), tricin (2), protocatechuic acid (3), luteolin (4), anemonin (5), scopoletin (6), 5-hydroxy-6, 7-dimethoxyflavone (7), 5-hydroxy-7, 8-dimethoxyflavone (8), ternatolide (9), respectively. Compounds 1-9 were isolated from R. japonicus for the first time while compounds 7, 8 were separated from Ranunculus genus first time.