MilliporeSigma
5'-methylaristeromycin and related derivatives.
The Journal of organic chemistry (2006-10-27)
Wei Ye, Stewart W Schneller
ABSTRACT
The biological versatility of aristeromycin (carbocyclic adenosine) is limited by accompanying cytotoxicity caused ostensibly by the intracellular formation of its 5'-nucleotide derivatives. Aristeromycin derivatives that offered steric interference to this transformation at the C-5' center were sought. This paper describes the facile stereospecific synthesis, where necessary, of such C-5'-methylated aristeromycin derivatives.