MilliporeSigma
  • Increased sensitivity of chitosan determination by a dye binding method.
Increased sensitivity of chitosan determination by a dye binding method.
Carbohydrate research (2006-11-08)
Christian Wischke, Hans-Hubert Borchert
ABSTRACT
Chitosan is a topic of current research in pharmaceutics, medicine, biotechnology, and beyond. This note describes an improved quantification of chitosan using the dye Cibacron Brilliant Red 3B-A. The method is sensitive and of a good reproducibility and linearity in the range of 10-80 microg/mL.