MilliporeSigma
  • Production of xylobiose from the autohydrolysis explosion liquor of corncob using Thermotoga maritima xylanase B (XynB) immobilized on nickel-chelated Eupergit C.
Production of xylobiose from the autohydrolysis explosion liquor of corncob using Thermotoga maritima xylanase B (XynB) immobilized on nickel-chelated Eupergit C.
Bioresource technology (2007-01-30)
S S Tan, D Y Li, Z Q Jiang, Y P Zhu, B Shi, L T Li
ABSTRACT
In this study, a thermostable recombinant xylanase B (XynB) from Thermotoga maritima MSB8 was immobilized on nickel-chelated Eupergit C 250L. This immobilized XynB was then used to hydrolyze the autohydrolysis explosion liquor of corncob (AELC) in a packed-bed enzyme reactor for continuous production of xylooligosaccharides, especially xylobiose. When tested in batch hydrolysis of AELC, the immobilized XynB still retained its relative activity of 92.5% after 10 cycles of hydrolysis at 90 degrees C. The immobilized XynB retained 83.6% of its initial hydrolysis activity even after 168 h of hydrolysis reaction at 90 degrees C and demonstrated a half-life time of 577.6 h (24 days) for continuous hydrolysis. HPLC showed that xylobiose (49.8%) and xylose (22.6%) were the main hydrolysis products yielded during continuous hydrolysis. Xylobiose was adsorbed on an activated charcoal column and eluted with a linear gradient of 15% (v/v) ethanol to yield xylobiose with 84.7% of recovery. Also, the purity of xylobiose was up to 97.2% as determined by HPLC. Therefore, the immobilized XynB was suitable for the efficient production of xylobiose from AELC. This is the first report on the immobilization of xylanase for xylobiose production.