MilliporeSigma
  • Endogenous corticosteroid biosynthesis in subjects after bilateral adrenalectomy.
Endogenous corticosteroid biosynthesis in subjects after bilateral adrenalectomy.
Clinical endocrinology (2007-03-27)
E M Freel, R Bernhardt, M Bernhardt, M Ingram, R Ingram, A M Wallace, R Fraser, E Davies, J M C Connell
ABSTRACT
Corticosteroids can be synthesized in extra-adrenal tissues but the contribution of this to circulating levels in humans is not known. Previous in vitro studies suggest that the 'hybrid' corticosteroid 18-oxocortisol (18-oxoF) is produced from cortisol by aldosterone synthase. We looked for evidence of extra-adrenal production of this and other corticosteroids in 10 subjects stable on long-term glucocorticoid replacement following bilateral adrenalectomy. In phase 1, patients were maintained on cortisol alone (30 mg/day), in phase 2 dexamethasone (2 mg/day), and in phase 3, both cortisol and dexamethasone. Each phase lasted 3 days. On the last day of each phase, 24-h urine collection was performed for analysis of steroid metabolite excretion [using gas chromatography-mass spectrometry (GCMS)] and plasma aldosterone and renin were measured (by radioimmunoassay). Cortisol metabolite excretion rate [tetrahydrocortisone (THE) + tetrahydrocortisol (THF) + allotetrahydrocortisol (aTHF)] fell from 9169 nmol/24 h in phase 1 to 22 nmol/24 h in phase 2, rising to 6843 nmol/24 h in phase 3. Tetrahydroaldosterone (THAldo) excretion was readily detectable and did not alter significantly between phases (26.5, 23.5 and 28.5 nmol/24 h, respectively; P = 0.474). 18-Hydroxycortisol (18-OHF) excretion was easily detectable in phases 1 and 3 (252.5 and 212 nmol/24 h), falling in phase 2 (12 nmol/24 h). 18-oxoF excretion rates were lower but followed a similar pattern (1.62, 0.085 and 1.785 nmol/24 h in phases 1, 2 and 3, respectively). Significant levels of adrenal steroids are found in adrenalectomized subjects. We speculate that this occurs at extra-adrenal sites or in residual adrenal cortex tissue in an ACTH-independent manner. Our data suggest that aldosterone synthase, acting on cortisol, is the source of 18-oxoF and 18-OHF in these subjects. Further studies of corticosteroid production within adrenalectomized subjects, looking for evidence of adrenal regrowth or residual adrenal tissue, are justified.