MilliporeSigma
  • Mini-dose of thalidomide for treatment of primary myelofibrosis. Report of a case with complete reversal of bone marrow fibrosis and splenomegaly.
Mini-dose of thalidomide for treatment of primary myelofibrosis. Report of a case with complete reversal of bone marrow fibrosis and splenomegaly.
Haematologica (2007-04-05)
Alain Berrebi, Edit Feldberg, Irena Spivak, Lev Shvidel
ABSTRACT
We report a patient with very advanced myelofibrosis and huge splenomegaly who showed a complete hematological response to low dose thalidomide with reversal of splenomegaly and bone narrow fibrosis after 30 months of the treatment.