MilliporeSigma
  • Bradykinin- and substance P-induced edema formation in the hamster cheek pouch is tyrosine kinase dependent.
Bradykinin- and substance P-induced edema formation in the hamster cheek pouch is tyrosine kinase dependent.
Journal of applied physiology (Bethesda, Md. : 1985) (2007-04-14)
Israel Rubinstein
ABSTRACT
The purpose of this study was to determine whether protein tyrosine kinase, a ubiquitous family of intracellular signaling enzymes that regulates endothelial cell function, modulates bradykinin- and substance P-induced increase in macromolecular efflux from the intact hamster cheek pouch microcirculation. Using intravital microscopy, I found that suffusion of bradykinin or substance P (each, 0.5 and 1.0 microM) onto the cheek pouch elicited significant, concentration-dependent leaky site formation and increase in clearance of fluorescein isothiocyanate-dextran (FITC-dextran; molecular mass, 70 kDa; P < 0.05). These responses were significantly attenuated by suffusion of genistein (1.0 microM) or tyrphostin 25 (10 microM), two structurally unrelated, nonspecific protein tyrosine kinase inhibitors (P < 0.05). Conceivably, the kinase(s) involved in this process could be agonist specific because genistein was more effective than tyrphostin 25 in attenuating bradykinin-induced responses while the opposite was observed with substance P. Both inhibitors had no significant effects on adenosine (0.5 M)-induced responses (P > 0.5). Collectively, these data suggest that the protein tyrosine kinase metabolic pathway modulates, in part, the edemagenic effects of bradykinin and substance P in the intact hamster cheek pouch microcirculation in a specific fashion.