MilliporeSigma
  • Plasma and urine pharmacokinetics of niacin and its metabolites from an extended-release niacin formulation.
Plasma and urine pharmacokinetics of niacin and its metabolites from an extended-release niacin formulation.
International journal of clinical pharmacology and therapeutics (2007-08-30)
R M Menon, M H Adams, M A González, D S Tolbert, J H Leu, E A Cefali
ABSTRACT
To characterize plasma and urine pharmacokinetics of niacin and its metabolites after oral administration of 2,000 mg of extended-release (ER) niacin in healthy male volunteers. Niacin ER was administered to 12 healthy male subjects following a low-fat snack. Plasma was collected for 12 h post dose and was analyzed for niacin, nicotinuric acid (NUA), nicotinamide (NAM) and nicotinamide-N-oxide (NNO). Urine was collected for 96 h post dose and analyzed for niacin and its metabolites, NUA, NAM, NNO, N-methylnicotinamide (MNA) and N-methyl-2-pyridone-5-carboxamide (2PY). Mean niacin Cmax and AUC(0-t) values were 9.3 microg/ml and 26.2 microg x h/ml and were the highest of all analytes measured. Peak niacin and NUA levels occurred at 4.6 h (median) while tmax for NAM and NNO were 8.6 and 11.1 h, respectively. The mean plasma terminal half-life for niacin (0.9 h) and NUA (1.3 h) was shorter as compared to NAM (4.3 h). Urine recovery of niacin and metabolites accounted for 69.5% of the administered dose; only 3.2% was excreted as niacin. The highest recovery was for 2PY (37.9%), followed by MNA (16.0%) and NUA (11.6%). Mean half-lives for 2PY and MNA calculated in urine were 12.6 and 12.8 h, respectively. Niacin was extensively metabolized following oral administration, and about 70% of the administered dose is recovered in urine in 96 h as niacin, NUA, MNA, NNO, NAM and 2PY. The plasma levels of the parent niacin were higher than its metabolites though only about 3% of the unchanged drug is recovered in urine.
Sigma-Aldrich
Nicotinuric acid