MilliporeSigma
  • Intrinsic fluorescence quenching of glutathione transferase pi by glutathione binding.
Intrinsic fluorescence quenching of glutathione transferase pi by glutathione binding.
The Italian journal of biochemistry (1991-09-01)
A M Caccuri, A Aceto, N Rosato, C Di Ilio, F Piemonte, G Federici
ABSTRACT
The binding of the GSH to the GSH transferase pi quenches the protein intrinsic fluorescence more than the binding of GS-Me. The calculated dissociation constants are 38.6 microM and 90.9 microM for GSH and GS-Me, respectively. From the reported data it is evident that the binding of GSH to GSH transferase pi quenches the intrinsic fluorescence with two different mechanisms. The first one is a conformational change induced by the binding of the GSH and it is present also with the GS-Me binding. A second proposed mechanism is a contact quenching between the sulphydryl GSH group and a tryptophan residue. This suggests that at least one of the tryptophan residues is located near the GSH binding site.
Sigma-Aldrich
S-Methylglutathione