MilliporeSigma
  • A simple diffuse reflectance Fourier transform infrared spectroscopic determination of sulfur dioxide in ambient air using potassium tetrachloromercurate as the absorbing reagent.
A simple diffuse reflectance Fourier transform infrared spectroscopic determination of sulfur dioxide in ambient air using potassium tetrachloromercurate as the absorbing reagent.
Journal of AOAC International (2007-09-01)
Santosh Kumar Verma, Manas Kanti Deb, Samir Bajpai, Yukio Suzuki, Shekhar Kumar Sinha, Yuan Gong
ABSTRACT
This paper presents development of a simple, rapid, and precise analytical method for determination of sulfur dioxide in ambient air by a gas to solid-phase conversion method. Sulfur dioxide is determined in the form of sulfite (SO3(2-)) because the absorbing reagent, potassium tetrachloromercurate (TCM), used in this method absorbs sulfur dioxide from the atmosphere in the form of sulfite. Determination of submicrogram levels of sulfur dioxide was based on the selection of a quantitative analytical peak at 495 cm(-1) among the 3 observed vibrational peaks for the dichlorosulfitomercurate complex formed after reaction of sulfur dioxide with TCM and measurement of absorbance using diffuse reflectance Fourier transform infrared spectroscopy. The limits of detection and quantification of the method were found to be 0.09 and 0.4 microg/g SO3(2-), respectively. The precision in terms of standard deviation and relative standard deviation (RSD) at a level of 2 microg SO3(2-)/10.1 g KBr for n = 10 was found to be 0.036 microg SO3(2-) and 1.8%, respectively. The RSD (n = 10) for determination of sulfur dioxide in ambient air was observed to be in the range of 2.7-4.2%. The method proposed is time saving and eliminates the slow and cumbersome steps of pH maintenance of the reaction mixture and color formation of the U.S. Environmental Protection Agency recommended West and Gaeke spectrophotometric method and other methods for quantitative determination of sulfur dioxide.