MilliporeSigma
  • Assessment of triethanolamine salicylate release from the dermatological bases and the commercial products.
Assessment of triethanolamine salicylate release from the dermatological bases and the commercial products.
Pharmaceutica acta Helvetiae (1991-01-01)
A Babar, P J Chickhale, F M Plakogiannis
ABSTRACT
Recently, triethanolamine salicylate (TEAS) is frequently being incorporated in several over-the-counter topical analgesic pharmaceutical products. Since the clinical efficacy of such dosage form depends upon the release of the active ingredient at the site of application, the present study was undertaken to study the in vitro release of the (TEAS) from commonly used ointment bases and two most popular commercial products in the U.S. market. Also, the effects of various penetration enhancers, such as, ethanol, propylene glycol, polyethylene glycol-400, dimethyl-sulfoxide (DMSO), polysorbate-80 and urea were evaluated. In general, the drug release from the experimental formulations was higher than the commercial products studied. The inclusion of the penetration enhancing ingredients increased the drug release from some of the formulations evaluated. The hydrophilic emulsion base with 10% ethanol exhibited the best in vitro drug release. The apparent viscosity profiles of the formulations showed no definite relationship with the amounts of drug release. However, significant differences in the (TEAS) release from the experimental formulations were observed.