MilliporeSigma
  • Heterotricyclic himbacine analogs as potent, orally active thrombin receptor (protease activated receptor-1) antagonists.
Heterotricyclic himbacine analogs as potent, orally active thrombin receptor (protease activated receptor-1) antagonists.
Journal of medicinal chemistry (2007-09-15)
Mariappan V Chelliah, Samuel Chackalamannil, Yan Xia, Keith Eagen, Martin C Clasby, Xiaobang Gao, William Greenlee, Ho-Sam Ahn, Jacqueline Agans-Fantuzzi, George Boykow, Yunsheng Hsieh, Matthew Bryant, Jairam Palamanda, Tze-Ming Chan, David Hesk, Madhu Chintala
ABSTRACT
Pursuing our earlier efforts in the himbacine-based thrombin receptor antagonist area, we have synthesized a series of compounds that incorporate heteroatoms in the C-ring of the tricyclic motif. This effort has resulted in the identification of several potent heterocyclic analogs with excellent affinity for the thrombin receptor. Several of these compounds demonstrated robust inhibition of platelet aggregation in an ex vivo model in cynomolgus monkeys following oral administration. A detailed profile of 28b, a benchmark compound in this series, with a Ki of 4.3 nM, is presented.