MilliporeSigma
Mechanism of decongestant activity of alpha 2-adrenoceptor agonists.
Pulmonary pharmacology & therapeutics (2007-09-18)
M R Corboz, M A Rivelli, G G Mingo, R L McLeod, L Varty, Y Jia, J A Hey
ABSTRACT
The vascular bed in nasal mucosa of different species, including human, is highly vascularized and an extensive sinusoidal network of large capacitance vessels is present deep within the submucosa. When this network of venous sinusoids is engorged with blood, the swollen mucosa reduces the size of the airway lumen and congestion ensues. Nasal vasculature tone is strongly influenced by the sympathetic nervous system and the only drugs approved specifically to relieve vascular nasal obstruction are alpha-adrenoceptor sympathomimetic agents. Due to their vasoconstrictor action, the sympathomimetic decongestants oppose vasodilation, reducing nasal airway resistance and thus facilitating nose breathing. However, standard decongestants that are non-selective alpha-adrenoceptor agonists are associated with the potential for side-effect liabilities including hypertension, stroke, insomnia and nervousness. We propose than a selective alpha 2-adrenoceptor agonist, by acting preferentially on nasal venous capacitance vessels, will elicit decongestion with a reduced side-effect liability. In the present study, we evaluated the effects of the selective alpha 2-adrenoceptor agonist BHT-920 in a real-time tissue contractility assay using isolated pig nasal explants and in an in vivo cat model of congestion. The vasoconstrictor and decongestant effects of BHT-920 were compared to the non-selective alpha-adrenoceptor agonist epinephrine and the standard decongestant oxymetazoline. Our results showed that the alpha 2-adrenoceptor agonist BHT-920 preferentially contracts venous sinusoids confirming previous observations [Corboz MR, Varty LM, Rivelli MA, Mutter JC, Mingo G, McLeod R, et al. Effects of an alpha 2-adrenoceptor agonist in nasal mucosa. Arch Physiol Biochem 2003;11: 335-6, Corboz MR, Rivelli MA, Varty LM, Mutter J, Cartwright M, Rizzo CA, et al. Pharmacological characterization of postjunctional alpha-adrenoceptor in human nasal mucosa. Am J Rhinol 2005;19: 495-502] and displays decongestion without affecting blood pressure. Therefore, an alpha 2-adrenoceptor agonist, by causing constriction in the capacitance vessels of nasal mucosa, can produce nasal decongestion without the effects on blood pressure observed with the standard selective alpha 1-adrenoceptor and non-selective alpha-adrenoceptor sympathomimetic decongestants.