MilliporeSigma
  • Synthesis, characterization and cytotoxicity of diam(m)-ineplatinum(II) complexes containing beta-phenylisosuccinate ligand.
Synthesis, characterization and cytotoxicity of diam(m)-ineplatinum(II) complexes containing beta-phenylisosuccinate ligand.
Archiv der Pharmazie (2007-10-10)
Qing-Song Ye, Li-Guang Lou, Zuo-Yan Liu, Wei-Ping Liu, Shu-Qian Hou, Xi-Zhu Chen, Yao Yu
ABSTRACT
Four diam(m)ineplatinum(II) complexes containing beta-phenylisosuccinate as the leaving groups were prepared, characterized, and evaluated for their cytotoxicity against A549/ATCC human lung cancer cell line and SGC-7901 human gastric cancer cell line. One of the complexes, (trans-1R,2R-diaminocyclohexane)-beta-phenylisosuccinatoplatinum(II) 4, was much more active than cisplatin and carboplatin.
Sigma-Aldrich
Phenylsuccinic acid, 98%