MilliporeSigma
  • Tungstate sulfuric acid (TSA)/KMnO4 as a novel heterogeneous system for rapid deoximation.
Tungstate sulfuric acid (TSA)/KMnO4 as a novel heterogeneous system for rapid deoximation.
Molecules (Basel, Switzerland) (2007-11-01)
Bahador Karami, Morteza Montazerozohori
ABSTRACT
Neat chlorosulfonic acid reacts with anhydrous sodium tungstate to give tungstate sulfuric acid (TSA), a new dibasic inorganic solid acid in which two sulfuric acid molecules connect to a tungstate moiety via a covalent bond. A variety of oximes were oxidized to their parent carbonyl compounds under mild conditions with excellent yields in short times by a heterogeneous wet TSA/KMnO4 in dichloromethane system.
Sigma-Aldrich
Acetaldehyde oxime, mixture of syn and anti, 99%
Pricing and availability is not currently available.