MilliporeSigma
  • A rapid and sensitive UPLC-ESI MS method for analysis of isofraxidin, a natural antistress compound, and its metabolites in rat plasma.
A rapid and sensitive UPLC-ESI MS method for analysis of isofraxidin, a natural antistress compound, and its metabolites in rat plasma.
Journal of separation science (2007-11-02)
Hui Sun, Haitao Lv, Yanmei Zhang, Xijun Wang, Kaishun Bi, Hongxin Cao
ABSTRACT
Isofraxidin is one of the main bioactive constituents in the root of Acanthopanax senticosus, which has antifatigue, antistress, and immuno-accommondating effects. In this study, an ultraperformance LC (UPLC)-ESI MS method was developed for analyzing isofraxidin and its metabolites in rat plasma. The analysis was performed on a UPLC coupled with ESI MS (quadropole MS tandem TOF MS). The lower LOD (LLOD) for isofraxidin was 0.25 ng/mL, the intraday precision was less than 10%, the interday precision was less than 10%, and the extraction recovery was more than 80%. Isofraxidin and two metabolites (M1 and M2) were detected in rat plasma after oral administration of isofraxidin, and the molecular polarities of M1 and M2 were both increased compared to isofraxidin. The metabolites were identified as 5,6-dihydroxyl-7-methoxycoumarin and 5-hydroxyl-6,7-dimethoxycoumarin when subjected to parent ion spectra, product ion spectra, and extract mass and element composition analyses.