MilliporeSigma
A loop resonator for slice-selective in vivo EPR imaging in rats.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2007-11-17)
Hiroshi Hirata, Guanglong He, Yuanmu Deng, Ildar Salikhov, Sergey Petryakov, Jay L Zweier
ABSTRACT
A loop resonator was developed for 300 MHz continuous-wave electron paramagnetic resonance (CW-EPR) spectroscopy and imaging in live rats. A single-turn loop (55 mm in diameter) was used to provide sufficient space for the rat body. Efficiency for generating a radiofrequency magnetic field of 38 microT/W(1/2) was achieved at the center of the loop. For the resonator itself, an unloaded quality factor of 430 was obtained. When a 350 g rat was placed in the resonator at the level of the lower abdomen, the quality factor decreased to 18. The sensitive volume in the loop was visualized with a bottle filled with an aqueous solution of the nitroxide spin probe 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy (3-CP). The resonator was shown to enable EPR imaging in live rats. Imaging was performed for 3-CP that had been infused intravenously into the rat and its distribution was visualized within the lower abdomen.