MilliporeSigma
  • Acetazolamide prevents vacuolar myopathy in skeletal muscle of K(+) -depleted rats.
Acetazolamide prevents vacuolar myopathy in skeletal muscle of K(+) -depleted rats.
British journal of pharmacology (2008-03-18)
D Tricarico, S Lovaglio, A Mele, G Rotondo, E Mancinelli, G Meola, D C Camerino
ABSTRACT
Acetazolamide and dichlorphenamide are carbonic anhydrase (CA) inhibitors effective in the clinical condition of hypokalemic periodic paralysis (hypoPP). Whether these drugs prevent vacuolar myopathy, which is a pathogenic factor in hypoPP, is unknown. The effects of these drugs on the efflux of lactate from skeletal muscle were also investigated. For 10 days, K(+)-depleted rats, a model of hypoPP, were administered 5.6 mg kg(-1) day(-1) of acetazolamide, dichlorphenamide or bendroflumethiazide (the last is not an inhibitor of CA). Histological analysis of vacuolar myopathy and in vitro lactate efflux measurements were performed in skeletal muscles from treated and untreated K(+)-depleted rats, and also from normokalemic rats. About three times as many vacuoles were found in the type II fibres of tibialis anterioris muscle sections from K(+)-depleted rats as were found in the same muscle from normokalemic rats. In ex vivo experiments, a higher efflux of lactate on in vitro incubation was found in muscles of K(+)-depleted rats compared with that found in muscles from normokalemic rats. After treatment of K(+)-depleted rats with acetazolamide, the numbers of vacuoles in tibialis anterioris muscle decreased to near normal values. Incubation with acetazolamide in vitro inhibited efflux of lactate from muscles of K(+)-depleted rats. In contrast, bendroflumethiazide and dichlorphenamide failed to prevent vacuolar myopathy after treatment in vivo and failed to inhibit lactate efflux in vitro. Acetazolamide prevents vacuolar myopathy in K(+)-depleted rats. This effect was associated with inhibition of lactate transport, rather than inhibition of CA.