MilliporeSigma
  • "Diversity oriented synthesis" of functionalized chiral tetrahydropyridines: potential GABA receptor agonists and azasugars from natural amino acids via a sequential Baylis-Hillman reaction and RCM protocol.
"Diversity oriented synthesis" of functionalized chiral tetrahydropyridines: potential GABA receptor agonists and azasugars from natural amino acids via a sequential Baylis-Hillman reaction and RCM protocol.
Journal of combinatorial chemistry (2008-04-16)
Palakodety Radha Krishna, P Srinivas Reddy
ABSTRACT
The preparation of chiral tetrahydropyridine-4-carboxylates as isoguvacine analogues and azasugars with a tertiary stereocenter from L-amino acids via diastereoselective a Baylis-Hillman reaction of N-allyl-Boc alpha-aminal, followed by ring-closing metathesis and dihydroxylation sequences, is reported.
Sigma-Aldrich
Isoguvacine hydrochloride, solid