MilliporeSigma
A practical approach to warts in the emergency department.
Pediatric emergency care (2008-04-18)
Bruce E Herman, Howard M Corneli
ABSTRACT
Although cutaneous warts are one of the most common skin conditions affecting children, their management can be challenging, especially in complex cases and even more so during a single emergency department encounter. Anogenital warts require particular attention because of their possible but nonspecific association with sexual abuse. This review will focus on cutaneous warts and anogenital warts, with a discussion of their recognition and treatment, especially in the emergency department. The medicolegal implications of anogenital warts in children are also reviewed.