MilliporeSigma
  • Analysis of triallate residues in cereals and soil by gas chromatography with ion-trap detection.
Analysis of triallate residues in cereals and soil by gas chromatography with ion-trap detection.
Journal of chromatography (1991-01-02)
C Sanchez-Brunete, T Salto, J M Garcia-Baudin, J L Tadeo
ABSTRACT
Triallate residues in barley seedlings and soil samples were determined by gas chromatography with ion-trap detection. Soil was extracted with methanol on a mechanical shaker, and plants were extracted with acetonitrile in a Sorvall homogenizer. After evaporation of the organic solvents, the residue was dissolved in hexane, and plants extracts were cleaned-up on an alumina column. Gas chromatographic analysis was carried out using a BP-1 fused-silica capillary column with helium as carrier gas. To quantitate residues the total-ion chromatogram was obtained and then the selected-ion monitoring chromatograms were displayed at m/z 86 for triallate and at m/z 154 for the internal standard, methyl-(4-amino-2-chloro)-benzoate. The average recovery through the method from barley and soil samples was always higher than 80%. The limit of detection in the selected-ion mode was 0.01 mg/kg. Barley and soil samples treated with triallate were also analysed. A good agreement was observed between results obtained by this method and by gas chromatography with nitrogen-phosphorus detection.
Product Description
Supelco
Triallat, PESTANAL®, analytical standard