MilliporeSigma
  • Simultaneous determination of p-arsanilic acid and roxarsone in feed by liquid chromatography-hydride generation online coupled with atomic fluorescence spectrometry.
Simultaneous determination of p-arsanilic acid and roxarsone in feed by liquid chromatography-hydride generation online coupled with atomic fluorescence spectrometry.
Journal of environmental monitoring : JEM (2008-08-09)
Jianjing Liu, Hongxia Yu, Haibin Song, Jing Qiu, Fengmei Sun, Ping Li, Shuming Yang
ABSTRACT
A novel, simple and sensitive liquid chromatography-hydride generation online coupled with atomic fluorescence spectrometry (LC-HG-AFS) method was developed for simultaneous determination of p-arsanilic acid (p-ASA) and roxarsone in feed. 20% Methanol aqueous was used as extraction reagent, after preprocessing samples by ultrasonic oscillation, then injected into the chromatography Waters symmetry shield RP18 analytical column (150mm x 4.6mm, 5 microm), finally detected by an atomic fluorescence spectrometer. The calibration curves of analyses were linear over a range of concentrations (0.2-4mg L-1 and the correlation coefficients were higher than 0.9990. The limits of detection were 0.2 mg L-1. The method has been validated by linearity, precision and recovery. p-ASA and roxarsone in feed can be successfully and simultaneously determined using the developed method without a tedious pretreatment procedure.