MilliporeSigma
  • Determination of Cd and Cr in an ABS candidate reference material by instrumental neutron activation analysis.
Determination of Cd and Cr in an ABS candidate reference material by instrumental neutron activation analysis.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2008-08-22)
Kwangwon Park, Namgoo Kang, Kyunghaeng Cho, Jounghae Lee
ABSTRACT
In order to practically better cope with technical barriers to trade (TBT) of a great number of resin goods, our research presents first-ever results for the determination of Cd and Cr in acrylonitrile butadiene styrene (ABS) candidate reference material using instrumental neutron activation analysis (INAA) recently recognized as a candidate primary ratio method with a particular attention to the estimation of involved measurement uncertainties.