MilliporeSigma
  • Application of an ovomucoid-conjugated polymer column for the enantiospecific determination of chlorprenaline concentrations in plasma.
Application of an ovomucoid-conjugated polymer column for the enantiospecific determination of chlorprenaline concentrations in plasma.
Journal of chromatography (1991-05-03)
T Miwa, S Sakashita, H Ozawa, J Haginaka, N Asakawa, Y Miyake
ABSTRACT
An ovomucoid-conjugated polymer column was prepared for the liquid chromatographic resolution of racemic compounds. The column showed strong retention of acidic solutes, a characteristic attributed to the structure of the stationary phase support gel. Although the efficiency of the column was lower than that of an ovomucoid-conjugated silica gel column, enantiospecific chlorprenaline determination in plasma was achieved with solute amounts from 1.0 ng to 0.1 microgram.