MilliporeSigma
  • Sigma1 receptor antagonists determine the behavioral pattern of the methamphetamine-induced stereotypy in mice.
Sigma1 receptor antagonists determine the behavioral pattern of the methamphetamine-induced stereotypy in mice.
Psychopharmacology (2008-12-05)
J Kitanaka, N Kitanaka, T Tatsuta, F S Hall, G R Uhl, K Tanaka, N Nishiyama, Y Morita, M Takemura
ABSTRACT
The effects of sigma receptor antagonists on methamphetamine (METH)-induced stereotypy have not been examined. We examined the effects of sigma antagonists on METH-induced stereotypy in mice. The administration of METH (10 mg/kg) to male ddY mice induced stereotyped behavior consisting of biting (90.1%), sniffing (4.2%), head bobbing (4.1%), and circling (1.7%) during an observation period of 1 h. Pretreatment of the mice with BMY 14802 (alpha-(4-fluorophenyl)-4-(5-fluoro-2-pyrimidinyl)-1-piperazinebutanol; 1, 5, and 10 mg/kg), a non-specific sigma receptor antagonist, significantly increased METH-induced sniffing (19.2%, 30.5%, and 43.8% of total stereotypical behavior) but decreased biting (76.6%, 66.9%, and 49.3% of total stereotypical behavior) in a dose-dependent manner. This response was completely abolished by (+)-SKF 10,047 ([2S-(2alpha,6alpha,11R)]-1,2,3,4,5,6-hexahydro-6,11-dimethyl-3-(2-propenyl)-2,6-methano-3-benzazocin-8-ol; 4 and 10 mg/kg), a putative sigma(1) receptor agonist, and partially by PB 28 (1-cyclohexyl-4-[3-(1,2,3,4-tetrahydro-5-methoxy-1-naphthalen-1-yl)-n-propyl]piperazine; 1 and 10 mg/kg), a putative sigma(2) receptor agonist. The BMY 14802 action on METH-induced stereotypy was mimicked by BD 1047 (N-[2-(3,4-dichlorophenyl)ethyl]-N-methyl-2-(dimethylamino)ethylamine; 10 mg/kg), a putative sigma(1) receptor antagonist, but not by SM-21 ((+/-)-tropanyl 2-(4-chlorophenoxy)butanoate; 1 mg/kg), a putative sigma(2) receptor antagonist. The BD 1047 effect on METH-induced stereotypy was also abolished completely by (+)-SKF 10,047 and partially by PB 28. The overall frequency of METH-induced stereotypical behavior was unchanged with these sigma receptor ligands, despite the alteration in particular behavioral patterns. The BMY 14802 action on METH-induced stereotypy was unaffected by pretreatment with centrally acting histamine H(1) receptor antagonists (pyrilamine or ketotifen, 10 mg/kg), suggesting that these effects are independent of histamine H(1) receptor signaling systems. In summary, modulation of central sigma(1) receptors alters the pattern of METH-induced stereotypy, producing a shift from stereotypical biting to stereotypical sniffing, without affecting the overall frequency of stereotypical behavior.