MilliporeSigma
  • Involvement of local serotonin-2A but not serotonin-1B receptors in the reinforcing effects of ethanol within the posterior ventral tegmental area of female Wistar rats.
Involvement of local serotonin-2A but not serotonin-1B receptors in the reinforcing effects of ethanol within the posterior ventral tegmental area of female Wistar rats.
Psychopharmacology (2009-01-24)
Zheng-Ming Ding, Jamie E Toalston, Scott M Oster, William J McBride, Zachary A Rodd
ABSTRACT
Previous studies indicated that ethanol could be self-infused into the posterior ventral tegmental area (p-VTA) and that activation of local serotonin-3 (5-HT(3)) receptors was involved. 5-HT(1B) and 5-HT(2A) receptors are involved in the effects of 5-HT and ethanol on VTA dopamine neurons. The current study used the intracranial self-administration (ICSA) procedure to determine the involvement of local 5-HT(1B) and 5-HT(2A) receptors in the self-infusion of ethanol into the p-VTA. Female Wistar rats were implanted unilaterally with a guide cannula aimed at the p-VTA. Seven days after surgery, rats were placed into the two-lever operant conditioning chambers for ICSA tests. The tests consisted of four acquisition sessions with self-infusion of 200 mg% ethanol alone, two or three sessions with co-infusion of the 5-HT(1B) antagonist GR 55562 (10, 100, or 200 microM) or the 5-HT(2A) antagonist R-96544 (10, 100, or 200 microM) with 200 mg% ethanol, and one final session with 200 mg% ethanol alone. During the acquisition sessions, all rats readily self-infused ethanol and discriminated the active from inactive lever. Co-infusion of GR 55562, at all three doses, had no effect on the self-infusion of ethanol. In contrast, co-infusion of R-96544, at the two higher doses, attenuated responding on the active lever for ethanol infusion (p < 0.05). The results suggest that the reinforcing effects of ethanol within the p-VTA are modulated, at least in part, by activation of local 5-HT(2A), but not 5-HT(1B), receptors.