MilliporeSigma
[Studies on phenylpropanoids from herbs of Eriophyton wallichii].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2009-02-17)
Qiu-ling Fan, Jing Liu, Ming-ming Zhao, Fu-sen Han, Chang-heng Tan, Cai-guo Huang, Da-yuan Zhu
ABSTRACT
To study the chemical constituents of Eriophyton wallichii. Compounds were separated and purified by column chromatographic methods, and their structures were elucidated by spectroscopic methods. Eight phenylpropanoids were isolated and identified as martynoside (1), leucosceptoside A (2), citrusin B (3), (+)-dehydrodiconiferyl alcohol-4, 9-beta-D-glucopyranoside (4), liriodendrin (5), velutinoside 11[ (6), jionoside B, (7), stachysoside D (8), respectively. The eight compounds were firstly isolated from E. wallichii.
Product Description
Supelco
Eleutheroside E, analytical standard