MilliporeSigma
Polyketide-chain branching by an enzymatic Michael addition.
Angewandte Chemie (International ed. in English) (2009-03-07)
Björn Kusebauch, Benjamin Busch, Kirstin Scherlach, Martin Roth, Christian Hertweck
ABSTRACT
A new "branch" for polyketide synthases was discovered in the biosynthesis of the antimitotic rhizoxin complex in the endofungal bacterium Burkholderia rhizoxinica. Genetic engineering and the structural elucidation of pathway intermediates revealed that a complex polyketide chain is branched at the beta position by an unprecedented conjugate addition of an acetyl building block to an acryloyl precursor (see scheme).