MilliporeSigma
Protective effects of a coumarin derivative in diabetic rats.
Investigative ophthalmology & visual science (2009-03-13)
Claudio Bucolo, Keith W Ward, Emanuela Mazzon, Salvatore Cuzzocrea, Filippo Drago
ABSTRACT
Retinal microvascular cells play a crucial role in the pathogenesis of diabetic retinopathy. The endothelial effects of cloricromene, a novel coumarin derivative, on diabetic retinopathy induced by streptozotocin (STZ) in the rat were investigated. Cloricromene (10 mg/kg intraperitoneally) was administered daily in diabetic rats, and 60 days later eyes were enucleated for localization of nitrotyrosine, ICAM-1, VEGF, ZO-1, occludin, claudin-5, and VE-cadherin by immunohistochemical analysis. The effect of treatment was also evaluated by TNFalpha, ICAM-1, VEGF, and eNOS protein levels measurement in the retina with the respective ELISA kits. Blood-retinal barrier (BRB) integrity was also evaluated by Evans blue. Increased amounts of cytokines, adhesion molecule, and nitric oxide synthase were observed in retina. Cloricromene treatment significantly lowered retinal TNFalpha, ICAM-1, VEGF, and eNOS. Furthermore, immunohistochemical analysis for VEGF, ICAM-1, nitrotyrosine (a marker of peroxynitrite), and tight junctions revealed positive staining in the retina from STZ-treated rats. The degree of staining for VEGF, ICAM-1, nitrotyrosine, and tight junctions was markedly reduced in tissue sections obtained from diabetic rats treated with cloricromene. Treatment with cloricromene suppressed diabetes-related BRB breakdown by 45%. This study provides the first evidence that the new coumarin derivative cloricromene attenuates the degree of inflammation preserving the BRB in diabetic rats.