MilliporeSigma
  • Accidental ingestion of dosulepin presenting as atrial flutter in a child.
Accidental ingestion of dosulepin presenting as atrial flutter in a child.
European journal of pediatrics (2009-04-07)
Santabhanu Chakrabarti, James Fraser, Beverly Tsai-Goodman
ABSTRACT
This case highlights the importance of considering drug ingestion when the clinical features at presentation are unusual. To our knowledge, this is the first report of atrial flutter secondary to tricyclic antidepressant (TCA) overdose in a child.