MilliporeSigma
  • Fustin flavonoid attenuates beta-amyloid (1-42)-induced learning impairment.
Fustin flavonoid attenuates beta-amyloid (1-42)-induced learning impairment.
Journal of neuroscience research (2009-06-18)
Chun-Hui Jin, Eun-Joo Shin, Jae-Bong Park, Choon-Gon Jang, Zhengyi Li, Min Soo Kim, Kyo Hwan Koo, Hyoung-Jong Yoon, Sang-Jae Park, Won-Cheol Choi, Kiyofumi Yamada, Toshitaka Nabeshima, Hyoung-Chun Kim
ABSTRACT
Natural flavonoids ameliorate amyloid-beta peptide (Abeta)-induced neurotoxicity. We examined whether the fustin flavonoid affects Abeta-induced learning impairment in mice. Repeated treatment with fustin significantly attenuated Abeta (1-42)-induced conditioned fear and passive avoidance behaviors. This effect was comparable to that of EGb761, a standard extract of ginkgo. Fustin treatment significantly prevented decreases in acetylcholine (ACh) levels, choline acetyltransferase (ChAT) activity, and ChAT gene expression induced by Abeta (1-42). Fustin also consistently suppressed increases in acetyl cholinesterase (AChE) activity and AChE gene expression induced by Abeta (1-42). In addition, fustin significantly attenuated Abeta (1-42)-induced selective decreases in muscarinic M1 receptor gene expression and muscarinic M1 receptor binding activity (as determined by [(3)H]pirenzepine binding) by modulating extracellular signal-regulated kinase 1/2 (ERK 1/2) and cAMP response-element binding protein (CREB) phosphorylation and brain-derived neurotrophic factor (BDNF) expression. These effects of fustin were reversed by treatment with dicyclomine, a muscarinic M1 receptor antagonist, and SL327, a selective ERK inhibitor, but not by chelerythrine, a pan-protein kinase C (PKC) inhibitor. Taken together, our results suggest that fustin attenuates Abeta (1-42)-impaired learning, and that the ERK/CREB/BDNF pathway is important for the M1 receptor-mediated cognition-enhancing effects of fustin.
Sigma-Aldrich
Dicyclomine hydrochloride, ≥99% (TLC), powder